6-Amino-D-luciferin ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: (S)-2-(6-Amino-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid, (S)-2-(6-Amino-2-benzothiazolyl)-4,5-dihydro-4-thiazolecarboxylic acid, 6′-Amino-D-luciferin

A275680-10MG A275680-25MG A275680-50MG A275680-5MG
ALA275680-10MGEA 226.01 USD
ALA275680-10MG ALA275680-25MG ALA275680-50MG ALA275680-5MG
6-Amino-D-luciferin ≥98%
6-Amino-D-luciferin
Formula: C₁₁H₉N₃O₂S₂
Storage Temperature: Ambient
CAS Number: 161055-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85d7fd4857403e1df72eb7a12326fc96d322b998&vid=85d7fd4857403e1df72eb7a12326fc96d322b998&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR