Aristolochic acid C ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 10-Hydroxy-6-nitronaphtho[2,1-g][1,3]benzodioxole-5-carboxylicacid

A275388-100MG A275388-1MG A275388-25MG
ALA275388100MGEA 9404.91 USD
ALA275388100MG ALA275388-1MG ALA275388-25MG
Aristolochic acid C ≥95%
Aristolochic acid C

Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₁₆H₉NO₇
Storage Temperature: Freezer
CAS Number: 4849-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0a4d4213abc79b62f0d974d2d192b46dfc64f00&vid=f0a4d4213abc79b62f0d974d2d192b46dfc64f00&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR