AF1219 TFA ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC
Synonyms:
AF 12198
A275277-10MG
A275277-1MG
A275277-25MG
A275277-50MG
A275277-5MG
ALA275277-10MGEA
2573.46
USD
ALA275277-10MG
ALA275277-1MG
ALA275277-25MG
ALA275277-50MG
ALA275277-5MG
AF1219 TFA ≥98%, Moligand™
AF1219 TFA
Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.
Formula:
C₉₆H₁₂₃N₁₉O₂₂ (free base) Storage Temperature: Freezer |
CAS Number:
185413-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...