N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide ≥97%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
N-[[4-(1,4,8,11-Tetraazacyclotetradec-1-ylmethyl)phenyl]methyl]-2-pyridinemethanaminehexahydrobromide, AMD 3465 6HBr, AMD 3465 hexahydrobromide, GENZ-644494 hexahydrobromide
A274943-100MG
A274943-10MG
A274943-25MG
A274943-2MG
A274943-50MG
A274943-5MG
ALA274943100MGEA
495.81
USD
ALA274943100MG
ALA274943-10MG
ALA274943-25MG
ALA274943-2MG
ALA274943-50MG
ALA274943-5MG
N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide ≥97%
N-(4-((1,4,8,11-Tetraazacyclotetradecan-1-yl)methyl)benzyl)-1-(pyridin-2-yl)methanamine hexahydrobromide
Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.
Formula:
C₂₄H₄₄Br₆N₆ MW: 896.07 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD20926342 CAS Number: 185991-07-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...