A83-01 ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-(6-Methylpyridin-2-yl)-N-phenyl-4-(quinolin-4-yl)-1H-pyrazole-1-carbothioamide, A 83-01
A274862-10MG
A274862-1MG
A274862-25MG
A274862-50MG
A274862-5MG
ALA274862-10MGEA
56.91
USD
ALA274862-10MG
ALA274862-1MG
ALA274862-25MG
ALA274862-50MG
ALA274862-5MG
A83-01 ≥98%
A83-01
A 83-01 is a potent inhibitor of TGF-β type I receptor ALK5 kinase, type I nodal receptor ALK4 and type I nodal receptor ALK7, with IC50s of 12 nM, 45 nM and 7.5 nM against the transcription induced by ALK5, ALK4 and ALK7, respectively。
Formula:
C₂₅H₁₉N₅S MW: 421.516 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08705403 CAS Number: 909910-43-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...