Ac-Glu(OtBu)-OH ≥98%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
N-Acetyl-L-glutamic acid 5-tert-butyl ester
A195245-1G
A195245-250MG
A195245-5G
ALA195245-1GEA
46.46
USD
ALA195245-1G
ALA195245250MG
ALA195245-5G
Ac-Glu(OtBu)-OH ≥98%
Ac-Glu(OtBu)-OH
Formula:
C₁₁H₁₉NO₅ MW: 245.275 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236754 CAS Number: 84192-88-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...