3-Amino-5-methylphenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A194971-10G A194971-1G A194971-250MG A194971-5G
ALA194971-10GEA 698.16 USD
ALA194971-10G ALA194971-1G ALA194971250MG ALA194971-5G
3-Amino-5-methylphenol ≥98%
3-Amino-5-methylphenol
Formula: C7H9NO
MW: 123.1500 g/mol
MDL Number: MFCD00086326
CAS Number: 76619-89-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06715e063a824e6c6afee347f135201331f407df&vid=06715e063a824e6c6afee347f135201331f407df&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR