4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥97%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide
A194927-1G
A194927-250MG
A194927-25G
A194927-5G
ALA194927-1GEA
134.81
USD
ALA194927-1G
ALA194927250MG
ALA194927-25G
ALA194927-5G
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide ≥97%
4-(4-Amino-3-fluorophenoxy)-N-methyl-2-pyridinecarboxamide
Formula:
C₁₃H₁₂FN₃O₂ MW: 261.26 g/mol Melting Pt: 143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09033852 CAS Number: 757251-39-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...