Acetazolamide ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetamido-5-sulphonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulphonamide, 2-Acetamido-5-sulfonamido-1,3,4-thiadiazole, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide

A194116-100G A194116-25G A194116-5G
ALA194116-100GEA 170.91 USD
ALA194116-100G ALA194116-25G ALA194116-5G
Acetazolamide ≥99%, Moligand™
Acetazolamide

Acetazolamide is a carbonic anhydrase (CA) IX inhibitor, which inhibits HCA IX with IC50 of 30 nm. diuretic.

Formula: C₄H₆N₄O₃S₂
MW: 222.24 g/mol
Melting Pt: 255°C
Storage Temperature: Ambient
MDL Number: MFCD00003105
CAS Number: 59-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e4a496ba665c9d95976fbfbc15e5f682ddbaa4&vid=91e4a496ba665c9d95976fbfbc15e5f682ddbaa4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR