Pseudothiohydantoin ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-aminothiazol-4(5H)-one, 2-Amino-4,5-dihydro-1,3-thiazol-4-one

A193919-1G A193919-250MG A193919-25G A193919-5G
ALA193919-1GEA 17.01 USD
ALA193919-1G ALA193919250MG ALA193919-25G ALA193919-5G
Pseudothiohydantoin ≥98%
Pseudothiohydantoin
Formula: C₃H₄N₂OS
MDL Number: MFCD00003186
CAS Number: 556-90-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ab1b57809c06e1632439c1493cedf0957984b83&vid=0ab1b57809c06e1632439c1493cedf0957984b83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR