4-Amino-5-methoxy-6-chloropyrimidine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
4-Amino-6-chloro-5-methoxypyrimidine
A193602-1G
A193602-25G
A193602-5G
ALA193602-1GEA
37.91
USD
ALA193602-1G
ALA193602-25G
ALA193602-5G
4-Amino-5-methoxy-6-chloropyrimidine ≥97%
4-Amino-5-methoxy-6-chloropyrimidine
Formula:
C₅H₆ClN₃O MW: 159.574 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07787635 CAS Number: 5018-41-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...