4-Amino-3,5-difluorobenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-amino-3,5-difluorobenzoicacid

A193595-100MG A193595-1G A193595-250MG
ALA193595100MGEA 13.21 USD
ALA193595100MG ALA193595-1G ALA193595250MG
4-Amino-3,5-difluorobenzoic acid ≥97%
4-Amino-3,5-difluorobenzoic acid
Formula: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD03407440
CAS Number: 500577-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cae176b98088049a82965c278fb8c88fdf238bac&vid=cae176b98088049a82965c278fb8c88fdf238bac&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR