5-Amino-2-methoxybenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A192044-25G A192044-5G A192044-1G A192044-250MG
ALA192044-25GEA 1083.86 USD
ALA192044-25G ALA192044-5G ALA192044-1G ALA192044250MG
5-Amino-2-methoxybenzonitrile ≥98%
5-Amino-2-methoxybenzonitrile
Formula: C₈H₈N₂O
MDL Number: MFCD00723841
CAS Number: 214623-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb8b00939c3a0e8eec053d948305aa2b680e51e9&vid=fb8b00939c3a0e8eec053d948305aa2b680e51e9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR