2-Amino-6-methylpyridin-3-ol ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-3-hydroxy-6-methylpyridine

A191936-100MG A191936-1G A191936-250MG A191936-5G
ALA191936100MGEA 56.91 USD
ALA191936100MG ALA191936-1G ALA191936250MG ALA191936-5G
2-Amino-6-methylpyridin-3-ol ≥97%
2-Amino-6-methylpyridin-3-ol
Formula: C₆H₈N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD09839245
CAS Number: 20348-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0989b3f523c21a0cb05dd6cbc3e467f965ffe944&vid=0989b3f523c21a0cb05dd6cbc3e467f965ffe944&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR