4-Amino-1,1,1-trifluorobut-3-en-2-one ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 4-Amino-2-oxo-1,1,1-trifluorobut-3-ene, 4-Amino-1,1,1-trifluoro-3-buten-2-one

A191726-1G A191726-250MG A191726-5G
ALA191726-1GEA 63.56 USD
ALA191726-1G ALA191726250MG ALA191726-5G
4-Amino-1,1,1-trifluorobut-3-en-2-one ≥95%
4-Amino-1,1,1-trifluorobut-3-en-2-one
Formula: C₄H₄F₃NO
MDL Number: MFCD01569588
CAS Number: 184848-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9e1b01bb54a9dd635ebe85f8e628cd85abbd4c8&vid=c9e1b01bb54a9dd635ebe85f8e628cd85abbd4c8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR