Acridin-4-ol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A191678-1G A191678-250MG A191678-5G
ALA191678-1GEA 141.46 USD
ALA191678-1G ALA191678250MG ALA191678-5G
Acridin-4-ol ≥98%
Acridin-4-ol
Formula: C₁₃H₉NO
Storage Temperature: Ambient
CAS Number: 18123-20-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=537bcd09956c508b69ecccbeeefaa1381026ed3e&vid=537bcd09956c508b69ecccbeeefaa1381026ed3e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR