6-Aminoquinazolin-4(3H)-one ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A191536-1G A191536-250MG
ALA191536-1GEA 376.11 USD
ALA191536-1G ALA191536250MG
6-Aminoquinazolin-4(3H)-one ≥97%
6-Aminoquinazolin-4(3H)-one
Formula: C₈H₇N₃O
MDL Number: MFCD01219542
CAS Number: 17329-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf0746298a261a19c314ea8b60de5fdf631346c4&vid=cf0746298a261a19c314ea8b60de5fdf631346c4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR