3-(3-Aminophenyl)propionic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-(3-Aminophenyl)propanoic acid

A191401-1G A191401-250MG A191401-25G A191401-5G
ALA191401-1GEA 17.96 USD
ALA191401-1G ALA191401250MG ALA191401-25G ALA191401-5G
3-(3-Aminophenyl)propionic acid ≥97%
3-(3-Aminophenyl)propionic acid

3-(3-Aminophenyl)propionic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 99-101°C
Storage Temperature: Refrigerator
MDL Number: MFCD00153889
CAS Number: 1664-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b465c183eaef6521eb2bb7295342fc3ac996db1&vid=2b465c183eaef6521eb2bb7295342fc3ac996db1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR