2-Acetoxybenzoic anhydride ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Aspirin Impurity F

A190994-100MG A190994-1G A190994-250MG A190994-25MG A190994-5G
ALA190994100MGEA 28.41 USD
ALA190994100MG ALA190994-1G ALA190994250MG ALA190994-25MG ALA190994-5G
2-Acetoxybenzoic anhydride ≥95%
2-Acetoxybenzoic anhydride
Formula: C₁₈H₁₄O₇
MW: 342.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00013236
CAS Number: 1466-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ac432426565f77e61ab9f0a538e819b77573f0d&vid=9ac432426565f77e61ab9f0a538e819b77573f0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR