Aluminum metaphosphate, Reagent Grade

Supplier: ALADDIN SCIENTIFIC
Warning

A190755-100G A190755-500G
ALA190755-100GEA 49.31 USD
ALA190755-100G ALA190755-500G
Aluminum metaphosphate, Reagent Grade
Aluminum metaphosphate
Formula: Al(PO₃)₃
Storage Temperature: Ambient
MDL Number: MFCD00075314
CAS Number: 32823-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6a4bf033bc74d6705aa9b71f2080c28ec5af123&vid=e6a4bf033bc74d6705aa9b71f2080c28ec5af123&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR