4-Amino-3,5-difluorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A189668-1G A189668-250MG A189668-25G A189668-5G
ALA189668-1GEA 39.81 USD
ALA189668-1G ALA189668250MG ALA189668-25G ALA189668-5G
4-Amino-3,5-difluorobenzonitrile ≥98%
4-Amino-3,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154.1169 g/mol
Melting Pt: 110-111°C
MDL Number: MFCD03407467
CAS Number: 110301-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86dc6dbdef7250792c11e78bb58526da52b9065d&vid=86dc6dbdef7250792c11e78bb58526da52b9065d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR