Aminoguanidine sulfate ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: Hydrazinecarboximidamide sulfate, 2-aminoguanidine, sulfuric acid, Guanylhydrazinesulphate, Guanylhydrazinesulfate, 2-aminoguanidine, sulphuric acid, Hydrazinecarboximidamide sulphate

A189564-250G A189564-25G
ALA189564-250GEA 165.21 USD
ALA189564-250G ALA189564-25G
Aminoguanidine sulfate ≥98%
Aminoguanidine sulfate
Formula: CH₈N₄O₄S
MDL Number: MFCD00151369
CAS Number: 1068-42-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c4405878140c5c64d032b179a0e01874d6b42bd&vid=7c4405878140c5c64d032b179a0e01874d6b42bd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR