6-Amino-4-hydroxy-2-mercaptopyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 6-Amino-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, 6-Aminothiouracil, 6-Amino-2-thiouracil

A189279-100G A189279-25G A189279-5G
ALA189279-100GEA 141.46 USD
ALA189279-100G ALA189279-25G ALA189279-5G
6-Amino-4-hydroxy-2-mercaptopyrimidine ≥98%
6-Amino-4-hydroxy-2-mercaptopyrimidine
Formula: C₄H₅N₃OS
MW: 143.1670 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006077
CAS Number: 1004-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba36c258dc7f572f1d5083358231e4bd9fdac1f0&vid=ba36c258dc7f572f1d5083358231e4bd9fdac1f0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR