2-Amidinopyrimidine hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A189006-1G A189006-25G A189006-5G
ALA189006-1GEA 11.31 USD
ALA189006-1G ALA189006-25G ALA189006-5G
2-Amidinopyrimidine hydrochloride ≥98%
2-Amidinopyrimidine hydrochloride
Formula: C₅H₇ClN₄
MW: 158.5889 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02677965
CAS Number: 138588-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7994a9d2f9c25d3131f7de9099a8bfae1d4a45b&vid=e7994a9d2f9c25d3131f7de9099a8bfae1d4a45b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR