(m-Acrylamidophenyl)boronic acid ≥97% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Acrylamidophenylboronic acid, (3-Acrylamidophenyl)boronic acid

A188951-1G A188951-250MG
ALA188951-1GEA 22.71 USD
ALA188951-1G ALA188951250MG
(m-Acrylamidophenyl)boronic acid ≥97% (contains varying amounts of Anhydride)
(m-Acrylamidophenyl)boronic acid

This acrylamide monomer contains a glucose sensitive boronic acid commonly used for intelligent polymers solutions and gels;for drug delivery systems;sensors;actuator systems;and bioreactors.

Formula: C₉H₁₀BNO₃
MW: 191 g/mol
Melting Pt: 129-146°C
Storage Temperature: Refrigerator
MDL Number: MFCD09025755
CAS Number: 99349-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf9ea84cd0dccec6d23ecc458e54e64c66fe443d&vid=bf9ea84cd0dccec6d23ecc458e54e64c66fe443d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR