2-Acetylbenzonitrile ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetylbenzenecarbonitrile

A188042-1G A188042-250MG
ALA188042-1GEA 44.56 USD
ALA188042-1G ALA188042250MG
2-Acetylbenzonitrile ≥95%
2-Acetylbenzonitrile
Formula: C₉H₇NO
Melting Pt: 46-48°C
Storage Temperature: Ambient
MDL Number: MFCD00272571
CAS Number: 91054-33-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4ece9da0cf25fc7bc015e4f9271ae4618e1bb0b&vid=e4ece9da0cf25fc7bc015e4f9271ae4618e1bb0b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR