3-Aminocyclopentanecarboxylic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A187884-100MG A187884-1G A187884-250MG A187884-5G
ALA187884100MGEA 36.01 USD
ALA187884100MG ALA187884-1G ALA187884250MG ALA187884-5G
3-Aminocyclopentanecarboxylic acid ≥95%
3-Aminocyclopentanecarboxylic acid
Formula: C₆H₁₁NO₂
MW: 129.15 g/mol
Boiling Pt: 264.7°C
Storage Temperature: Ambient
MDL Number: MFCD01318271
CAS Number: 89614-96-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf26517db6943b44385694c0a97897d40a7b7908&vid=cf26517db6943b44385694c0a97897d40a7b7908&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR