2-Aminocyclopentanol ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Amino cyclopentanol

A187867-10G A187867-1G A187867-25G A187867-5G
ALA187867-10GEA 223.16 USD
ALA187867-10G ALA187867-1G ALA187867-25G ALA187867-5G
2-Aminocyclopentanol ≥98%
2-Aminocyclopentanol
Formula: C₅H₁₁NO
MW: 101.1469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00100903
CAS Number: 89381-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b3460a8a254cabaaa767f1194c6a7ef1f8f6f81&vid=4b3460a8a254cabaaa767f1194c6a7ef1f8f6f81&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR