5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Bromo-4,5-(difluoromethylenedioxy)aniline, 6-Bromo-2,2-difluorobenzo[d][1,3]dioxol-5-amine, 6-Bromo-2,2-difluoro-1,3-benzodioxol-5-amine

A187780-10G A187780-1G A187780-250MG A187780-5G
ALA187780-10GEA 467.31 USD
ALA187780-10G ALA187780-1G ALA187780250MG ALA187780-5G
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole ≥95%
5-Amino-6-bromo-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₄BrF₂NO₂
MW: 252.013 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972651
CAS Number: 887267-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4baa7fc40c66cdb01cfc1321d11997d2562fcc5&vid=b4baa7fc40c66cdb01cfc1321d11997d2562fcc5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR