2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxamide

A186586-10G A186586-1G A186586-250MG A186586-5G
ALA186586-10GEA 429.31 USD
ALA186586-10G ALA186586-1G ALA186586250MG ALA186586-5G
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide ≥95%
2-Amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid amide
Formula: C₈H₁₀N₂OS
MW: 182.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00122463
CAS Number: 77651-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dbb99f9b6356f353705ab660cbadfb3456e9a0f&vid=7dbb99f9b6356f353705ab660cbadfb3456e9a0f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR