5-Acetyl-2-fluorobenzeneboronic acid pinacol ester ≥96%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-(4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 5-Acetyl-2-fluorophenylboronic acid pinacol ester
A186494-250MG
A186494-5G
A186494-1G
ALA186494250MGEA
70.21
USD
ALA186494250MG
ALA186494-5G
ALA186494-1G
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester ≥96%
5-Acetyl-2-fluorobenzeneboronic acid pinacol ester
Formula:
C₁₄H₁₈BFO₃ MW: 264.103 g/mol Melting Pt: 53-55°C Storage Temperature: Ambient |
MDL Number:
MFCD16996292 CAS Number: 765916-70-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...