3-Bromo-5-chloropyrazin-2-amine ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Amino-3-bromo-5-chloropyrazine

A186489-1G A186489-5G
ALA186489-1GEA 19.86 USD
ALA186489-1G ALA186489-5G
3-Bromo-5-chloropyrazin-2-amine ≥96%
3-Bromo-5-chloropyrazin-2-amine
Formula: C4H3BrClN3
MW: 208.4400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09909643
CAS Number: 76537-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2df58c53a1af70185332f3be1900807670c029bb&vid=2df58c53a1af70185332f3be1900807670c029bb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR