6-Bromo-2-naphthylamine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
6-Bromonaphthalen-2-amine, 2-Amino-6-bromonaphthalene
A186412-100G
A186412-1G
A186412-250MG
A186412-25G
A186412-5G
ALA186412-100GEA
553.76
USD
ALA186412-100G
ALA186412-1G
ALA186412250MG
ALA186412-25G
ALA186412-5G
6-Bromo-2-naphthylamine ≥98%
6-Bromo-2-naphthylamine
Formula:
C₁₀H₈BrN MW: 222.0812 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01026466 CAS Number: 7499-66-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...