2-Amino-4-chloro-6-isopropylpyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Chloro-6-isopropylpyrimidin-2-amine

A186322-1G A186322-250MG A186322-5G
ALA186322-1GEA 221.26 USD
ALA186322-1G ALA186322250MG ALA186322-5G
2-Amino-4-chloro-6-isopropylpyrimidine ≥98%
2-Amino-4-chloro-6-isopropylpyrimidine

2-Amino-4-chloro-6-isopropylpyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Formula: C₇H₁₀ClN₃
Melting Pt: 98-101°C
Storage Temperature: Refrigerator
MDL Number: MFCD03840198
CAS Number: 73576-33-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7e5a84848cedc988236108b804dbaa25e1ccc5&vid=5e7e5a84848cedc988236108b804dbaa25e1ccc5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR