2-Aminopropanenitrile hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: rac-a-Aminopropionitrile Hydrochloride, 2-Aminopropanenitrile, HCl, α-DL-Alanylnitrile Hydrochloride, α-Aminopropionitrile Hydrochloride, DL-α-Aminopropionitrile Hydrochloride, Alaninenitrile Hydrochloride, 2-Aminopropanenitrile Hydrochloride

A186242-100MG A186242-1G A186242-250MG
ALA186242100MGEA 74.96 USD
ALA186242100MG ALA186242-1G ALA186242250MG
2-Aminopropanenitrile hydrochloride ≥97%
2-Aminopropanenitrile hydrochloride
Formula: C₃H₇ClN₂
MW: 106.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09971651
CAS Number: 72187-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34177d576d6509283c94e0da518b26011cd8c89a&vid=34177d576d6509283c94e0da518b26011cd8c89a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR