5-Nitro-1H-benzo[d]imidazol-2-amine ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-5-nitrobenzimidazole, 2-Amino-5-nitro-1h-benzimidazole, 6-Nitro-1H-benzo[d]imidazol-2-amine
A185619-1G
A185619-250MG
A185619-5G
ALA185619-1GEA
1007.86
USD
ALA185619-1G
ALA185619250MG
ALA185619-5G
5-Nitro-1H-benzo[d]imidazol-2-amine ≥97%
5-Nitro-1H-benzo[d]imidazol-2-amine
Formula:
C₇H₆N₄O₂ MW: 178.15 g/mol Melting Pt: 246 °C |
MDL Number:
MFCD06659794 CAS Number: 6232-92-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...