2-Amino-3-(trifluoromethyl)benzonitrile ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Amino-3-trifluoromethylbenzonitrile
A185375-1G
A185375-25G
A185375-5G
ALA185375-1GEA
34.11
USD
ALA185375-1G
ALA185375-25G
ALA185375-5G
2-Amino-3-(trifluoromethyl)benzonitrile ≥98%
2-Amino-3-(trifluoromethyl)benzonitrile
Formula:
C₈H₅F₃N₂ MW: 186.1339 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972665 CAS Number: 58458-14-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...