Ethyl-3-amino-3-iminopropanoate hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Ethyl 2-amidinoacetate hydrochloride

A185319-1G A185319-25G A185319-5G
ALA185319-1GEA 22.71 USD
ALA185319-1G ALA185319-25G ALA185319-5G
Ethyl-3-amino-3-iminopropanoate hydrochloride ≥97%
Ethyl-3-amino-3-iminopropanoate hydrochloride
Formula: C₅H₁₁ClN₂O₂
MW: 166.60 g/mol
Melting Pt: 105-108°C
Storage Temperature: Ambient
MDL Number: MFCD06797615
CAS Number: 57508-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc8e4840d771676689cdc612eec2cb3f7cdaac50&vid=cc8e4840d771676689cdc612eec2cb3f7cdaac50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR