Anethole trithione ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Felviten

A185022-1G A185022-25G A185022-5G
ALA185022-1GEA 15.11 USD
ALA185022-1G ALA185022-25G ALA185022-5G
Anethole trithione ≥95%
Anethole trithione
Formula: C₁₀H₈OS₃
MW: 240.3649 g/mol
Melting Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00129751
CAS Number: 532-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3952d089e06679d0ab985cfd2607cdf2b2074909&vid=3952d089e06679d0ab985cfd2607cdf2b2074909&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR