2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid amide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A184722-100G A184722-10G A184722-1G A184722-25G A184722-5G
ALA184722-100GEA 1697.56 USD
ALA184722-100G ALA184722-10G ALA184722-1G ALA184722-25G ALA184722-5G
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid amide ≥97%
2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid amide
Formula: C₉H₁₂N₂OS
Melting Pt: 195-197°C
MDL Number: MFCD00111277
CAS Number: 4815-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ff267e59304858831e68be2319511a4fe2d5d16&vid=7ff267e59304858831e68be2319511a4fe2d5d16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR