2-Amino-3-hydroxypyrazine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A184511-100MG A184511-1G A184511-250MG
ALA184511100MGEA 11.31 USD
ALA184511100MG ALA184511-1G ALA184511250MG
2-Amino-3-hydroxypyrazine ≥97%
2-Amino-3-hydroxypyrazine
Formula: C₄H₅N₃O
Storage Temperature: Ambient
MDL Number: MFCD01646052
CAS Number: 43029-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2678b43f5295c4d3f708ba29a7fcbb9a07a6bb02&vid=2678b43f5295c4d3f708ba29a7fcbb9a07a6bb02&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR