4-Amino-3-nitroquinoline ≥98%

Supplier: ALADDIN SCIENTIFIC

A184486-100MG A184486-1G A184486-250MG A184486-25G A184486-5G
ALA184486100MGEA 112.01 USD
ALA184486100MG ALA184486-1G ALA184486250MG ALA184486-25G ALA184486-5G
4-Amino-3-nitroquinoline ≥98%
4-Amino-3-nitroquinoline
Formula: C₉H₇N₃O₂
Storage Temperature: Ambient
CAS Number: 42606-33-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aee64e06f7f3816fafbadb33f8772cc4eb0c190a&vid=aee64e06f7f3816fafbadb33f8772cc4eb0c190a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR