2-Amino-2,3-dimethylbutyramide ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A184403-500G
A184403-5G
A184403-100G
A184403-25G
ALA184403-500GEA
1930.31
USD
ALA184403-500G
ALA184403-5G
ALA184403-100G
ALA184403-25G
2-Amino-2,3-dimethylbutyramide ≥95%
2-Amino-2,3-dimethylbutyramide
Formula:
C₆H₁₄N₂O Melting Pt: 76.0 - 80.0 °C Storage Temperature: Ambient |
MDL Number:
MFCD02682920 CAS Number: 40963-14-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...