2-Amino-5-thiazolecarboxylic acid ≥96%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Aminothiazole-5-carboxylic acid

A184341-1G A184341-5G
ALA184341-1GEA 9.41 USD
ALA184341-1G ALA184341-5G
2-Amino-5-thiazolecarboxylic acid ≥96%
2-Amino-5-thiazolecarboxylic acid
Formula: C₄H₄N₂O₂S
MDL Number: MFCD06203554
CAS Number: 40283-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0595aace841b711d4b77f231036140f34aec2568&vid=0595aace841b711d4b77f231036140f34aec2568&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR