4-Amino-3-phenylbutanoic acid hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A183566-100G A183566-1G A183566-25G A183566-5G
ALA183566-100GEA 979.36 USD
ALA183566-100G ALA183566-1G ALA183566-25G ALA183566-5G
4-Amino-3-phenylbutanoic acid hydrochloride ≥98%
4-Amino-3-phenylbutanoic acid hydrochloride
Formula: C₁₀H₁₃NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD00075860
CAS Number: 3060-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c1b52886e6e90d3e59a89f87cf32e889767cdcc&vid=0c1b52886e6e90d3e59a89f87cf32e889767cdcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR