2-Amino-6-bromophenol ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
3-Bromo-2-hydroxyaniline
A183405-10G
A183405-1G
A183405-25G
ALA183405-10GEA
79.71
USD
ALA183405-10G
ALA183405-1G
ALA183405-25G
2-Amino-6-bromophenol ≥97%
2-Amino-6-bromophenol
Formula:
C₆H₆BrNO MW: 188.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277300 CAS Number: 28165-50-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...