Isoquinoline-8-amine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
Isoquinolin-8-amine, 8-Aminoisoquinoline
A183056-1G
A183056-250MG
A183056-25G
A183056-5G
ALA183056-1GEA
39.81
USD
ALA183056-1G
ALA183056250MG
ALA183056-25G
ALA183056-5G
Isoquinoline-8-amine ≥98%
Isoquinoline-8-amine
Formula:
C₉H₈N₂ MW: 144.1700 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00179553 CAS Number: 23687-27-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...