3-Amino-4-pyridazinecarboxylic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A182743-1G A182743-250MG A182743-5G
ALA182743-1GEA 163.31 USD
ALA182743-1G ALA182743250MG ALA182743-5G
3-Amino-4-pyridazinecarboxylic acid ≥95%
3-Amino-4-pyridazinecarboxylic acid
Formula: C5H5N3O2
Storage Temperature: Refrigerator
MDL Number: MFCD01318258
CAS Number: 21141-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd93ada759f81fef34a3b97112af532bc64aaeb3&vid=bd93ada759f81fef34a3b97112af532bc64aaeb3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR