2-Amino-5-bromonicotinaldehyde ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A182672-1G A182672-250MG A182672-25G A182672-5G
ALA182672-1GEA 30.31 USD
ALA182672-1G ALA182672250MG ALA182672-25G ALA182672-5G
2-Amino-5-bromonicotinaldehyde ≥98%
2-Amino-5-bromonicotinaldehyde
Formula: C₆H₅BrN₂O
MDL Number: MFCD09607904
CAS Number: 206997-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60ab7a66560aa44a7262e7e34fc57143f3c596a1&vid=60ab7a66560aa44a7262e7e34fc57143f3c596a1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR