3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 3-(Carbamoylmethyl)-5-methylhexanoic acid

A182244-100G A182244-10G A182244-25G A182244-5G
ALA182244-100GEA 43.61 USD
ALA182244-100G ALA182244-10G ALA182244-25G ALA182244-5G
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid ≥98%
3-(2-Amino-2-oxoethyl)-5-methylhexanoic acid
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Melting Pt: 132-136°C
Storage Temperature: Ambient
MDL Number: MFCD08457698
CAS Number: 181289-15-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5289409133491d60900fe9abd868fb2719d3a38&vid=f5289409133491d60900fe9abd868fb2719d3a38&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR